Human Anti-Herpes simplex virus Type 1 and 2 IgG ELISA Kit (HSV1+2)
Catalog # ABCAAB108741-96
Supplier: Abcam
CAS Number:
Specifications
- Descrizione:Human Anti-Herpes simplex virus Type 1 and 2 IgG ELISA kit (HSV1+2)
- Test per kit:96 tests
Specifications
About this item
Human Anti-Herpes simplex virus Type 1 and 2 IgG ELISA kit (HSV1+2) is a Indirect ELISA kit for the measurement of Human Anti-Herpes simplex virus Type 1 and 2 IgG (HSV1+2) in Human in Citrate plasma, Heparin Plasma, serum samples.