Mouse CXCL15 ELISA Kit
Catalog # ABCAAB234567-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse CXCL15 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse CXCL15 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse CXCL15 with a sensitivity of 1,7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.