Human IL-1 Beta ELISPOT Kit
Catalog # ABCAAB62924-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-1 Beta ELISPOT kit
- Seq:1
- Species reactivity:Human
- Sample type:Suspension cells
- Form:Pre-coated
- Storage temperature:+4 ℃
- Processing time:Multiple steps
- Antigen symbol:IL-1 beta
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human IL-1 Beta ELISPOT kit is a ELISPOT kit for the sensitive detection of Human IL-1 Beta production in a single cell suspension.