Order Entry
Italy
ContactUsLinkComponent
Mouse HGF ELISA Kit
Mouse HGF ELISA Kit
Catalog # ABCAAB223862-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse HGF ELISA Kit
Catalog # ABCAAB223862-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Mouse HGF ELISA kit
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse HGF ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse HGF with a sensitivity of 8,4 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.