Human NRG1 beta 1 ELISA Kit
Catalog # ABCAAB100614-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:NRG1 beta 1
- Description:Human NRG1 beta 1 ELISA kit
- Sample type:Plasma, cell culture supernatant
- Detection method:Colorimetric
- Detection range:82,3 - 20000 pg/ml
- Storage temperature:4 °C
- Sensitivity:50 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human NRG1 beta 1 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human NRG1 beta 1 in Human in plasma, cell culture supernatant samples.