Human IL-17F ELISPOT Kit (with un-coated plates)
Supplier: Abcam
Certificates
About this item
Human IL-17F ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Human IL-17F production in a single cell suspension.
Specifications
- Species reactivity:Species independent
- Sample type:Suspension cells
- Form:Uncoated
- Antigen name:IL-17F
- Storage temperature:+4 °C
- Processing time:Multiple steps
- Regulatory status:RUO
Specifications
Frequently Bought Together