Order Entry
Italy
ContactUsLinkComponent
Mouse Osteoprotegerin ELISA Kit (TNFRSF11B)
Mouse Osteoprotegerin ELISA Kit (TNFRSF11B)
Catalog # ABCAAB203365-96
Nome del fornitore:  Abcam
Numero CAS:  
undefined
Mouse Osteoprotegerin ELISA Kit (TNFRSF11B)
Catalog # ABCAAB203365-96
Nome del fornitore:  Abcam
Numero CAS:  

Specifiche

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    Osteoprotegerin
  • Description:
    Mouse Osteoprotegerin ELISA kit (TNFRSF11B)
  • Sample type:
    Heparin plasma, cell culture supernatant, serum
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Detection range:
    125 - 8000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    5 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifiche

Informazioni su questo articolo

Mouse Osteoprotegerin ELISA kit (TNFRSF11B) is a single-wash 90-min Simplestep used to quantify Mouse Osteoprotegerin (TNFRSF11B) with a sensitivity of 5 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.