STAT1 (pS727) ELISA Kit
Catalog # ABCAAB126454-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:STAT1
- Description:STAT1 (pS727) ELISA kit
- Sample type:Cell lysate
- Time to Results:5 h 0 min
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
STAT1 (pS727) ELISA kit is a Semi-quantitative ELISA kit for the measurement of STAT1 (pS727) in Mouse, Human in Cell Lysate samples.