STAT3 (Tyr705) In-Cell ELISA Kit
Catalog # ABCAAB126427-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:STAT3
- Description:STAT3 (Tyr705) In-Cell ELISA kit
- Sample type:Adherent cells
- Time to Results:5 h 10 min
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
STAT3 (Tyr705) In-Cell ELISA kit is Cell-based (qualitative) ELISA for the measurement of STAT3 (Tyr705) production in Mouse, Rat, Human in Adherent cells samples.