Human Anti-Chlamydia trachomatis IgM ELISA Kit
Catalog # ABCAAB108722-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Anti-Chlamydia trachomatis IgM ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human Anti-Chlamydia trachomatis IgM ELISA Kit is a Indirect ELISA kit for the measurement of Human Anti-Chlamydia trachomatis IgM in Human in Citrate plasma, Heparin Plasma, Serum samples.