Order Entry
Italy
ContactUsLinkComponent
Mouse Alpha-synuclein ELISA Kit
Mouse Alpha-synuclein ELISA Kit
Catalog # ABCAAB282865-1X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse Alpha-synuclein ELISA Kit
Catalog # ABCAAB282865-1X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    Alpha-synuclein
  • Description:
    Mouse Alpha-synuclein ELISA kit
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Detection range:
    11,7 - 750 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    7,2 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse Alpha-synuclein ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Alpha-synuclein with a sensitivity of 7.2 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.