Anti-GPCR GPRC5D Rabbit Monoclonal Antibody [clone: EPR28376-41]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR28376-41] to GPCR GPRC5D - BSA and Azide free.
Type: Primary
Antigen: GPCR GPRC5D
Clonality: Monoclonal
Clone: EPR28376-41
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together