Anti-CD8 alpha Rabbit Recombinant Multiclonal Antibody [clone: RM1129]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit recombinant multiclonal [RM1129] to CD8 alpha.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD8 alpha
Clonality: Recombinant Multiclonal
Clone: RM1129
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human
Specifications
- Catalog No:
- ABCAAB316778-100UL
- ABCAAB316778-20UL
- ABCAAB316778-1ML
- Antigen symbol:
- CD8 alpha
- CD8 alpha
- CD8 alpha
- Antigen name:
- CD8a
- CD8a
- CD8a
- Antigen synonyms:
- MAL|T-cell surface glycoprotein CD8 alpha chain|T-lymphocyte differentiation antigen T8/Leu-2
- MAL|T-cell surface glycoprotein CD8 alpha chain|T-lymphocyte differentiation antigen T8/Leu-2
- MAL|T-cell surface glycoprotein CD8 alpha chain|T-lymphocyte differentiation antigen T8/Leu-2
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Recombinant Multiclonal
- Recombinant Multiclonal
- Recombinant Multiclonal
- Clone:
- RM1129
- RM1129
- RM1129
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Liquid
- Molecular weight:
- 25729 Da
- 25729 Da
- 25729 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Shipping temperature:
- 4 °C
- 4 °C
- 4 °C
- Tested applications:
- Flow Cyt, Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt, Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt, Flow Cyt (Intra), ICC/IF, IHC-P
Specifications
Frequently Bought Together