Anti-RAGE Rabbit Recombinant Multiclonal Antibody
: Abcam
New Product
Rabbit recombinant multiclonal [RM1078] to RAGE.
Type: Primary
Antigen: RAGE
Clonality: Recombinant Multiclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together