Anti-Collagen I Rabbit Recombinant Multiclonal Antibody [clone: RM1131]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit recombinant multiclonal [RM1131] to Collagen I.
Type: Primary
Antigen: Collagen I
Clonality: Recombinant Multiclonal
Clone: RM1131
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together