Anti-IFIT5 Mouse Polyclonal Antibody
Product Details & Documents
Mouse Polyclonal IFIT5 antibody. Suitable for WB and reacts with Human samples. Immunogen corresponding to Recombinant Full Length Protein corresponding to Human IFIT5.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: IFIT5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Seq:1202
- Size:50 µg
- Pack type:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Protein A Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Host:Mouse
- Concentration:1 mg/ml
- Molecular weight:55847 Da
- Shipping temperature:4 °C
- Antigen name:ISG58
- Cat. No.:ABCAAB167272-50
- Reactivity:Human
- Antigen synonyms:RI58|Interferon-induced protein with tetratricopeptide repeats 5|IFIT-5|Interferon-induced 58 kDa protein|Retinoic acid- and interferon-inducible 58 kDa protein|P58
- Immunogen:Recombinant Full Length Protein corresponding to Human IFIT5
- Antigen symbol:IFIT5
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



