Anti-CD31 Rabbit Recombinant Multiclonal Antibody [clone: RM1006]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit recombinant multiclonal [RM1006] to CD31.
Type: Primary
Antigen: CD31
Clonality: Recombinant Multiclonal
Clone: RM1006
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together