Anti-SLC1A5/ASCT2 Rabbit Monoclonal Antibody [clone: CAL33]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [CAL33] to SLC1A5/ASCT2.
Type: Primary
Antigen: SLC1A5/ASCT2
Clonality: Monoclonal
Clone: CAL33
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together