Order Entry
Italy
ContactUsLinkComponent
Anti-Laminin beta 1 Rabbit Monoclonal Antibody [clone: EPR23265-32]
Anti-Laminin beta 1 Rabbit Monoclonal Antibody [clone: EPR23265-32]
Catalog # ABCAAB256380-1ML
Supplier:  Abcam
New Product
undefined
Anti-Laminin beta 1 Rabbit Monoclonal Antibody [clone: EPR23265-32]
Catalog # ABCAAB256380-1ML
Supplier:  Abcam
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Laminin subunit beta-1
  • Antigen symbol:
    Laminin beta 1
  • Clonality:
    Monoclonal
  • Clone:
    EPR23265-32
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    Laminin B1 chain|Laminin-1 subunit beta|Laminin-10 subunit beta|Laminin-12 subunit beta|Laminin-2 subunit beta|Laminin-6 subunit beta|Laminin-8 subunit beta|LAMB1
  • Molecular weight:
    198038 Da
  • Storage temperature:
    Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
  • Shipping temperature:
    4 °C
  • Immunogen:
    The exact immunogen used to generate this antibody is proprietary information.
  • Tested applications:
    ICC/IF
  • Purification:
    Protein A Affinity purification
  • Size:
    1 ml
  • Pack type:
    Vial

Specifications

About this item

Rabbit monoclonal [EPR23265-32] to Laminin beta 1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Laminin beta 1
Clonality: Monoclonal
Clone: EPR23265-32
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat