Order Entry
Italy
ContactUsLinkComponent
Anti-Insulin Receptor Rabbit Monoclonal Antibody [clone: Fab 83-7]
Anti-Insulin Receptor Rabbit Monoclonal Antibody [clone: Fab 83-7]
Catalog # ABCAAB245688-200
Supplier:  Abcam
undefined
Anti-Insulin Receptor Rabbit Monoclonal Antibody [clone: Fab 83-7]
Catalog # ABCAAB245688-200
Supplier:  Abcam

Specifications

  • Seq:
    1358
  • Size:
    200 µg
  • Pack type:
    Vial

Specifications

About this item

Rabbit monoclonal [Fab 83-7] to Insulin Receptor.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Insulin Receptor
Clonality: Monoclonal
Clone: Fab 83-7
Conjugation:
Epitope: This antibody recognizes an epitope within the cysteine rich region (amino acids 140-301) of the extracellular domain of the human insulin receptor alpha
Host: Rabbit
Isotype: IgG
Reactivity: Human