Anti-STAT1 Mouse Monoclonal Antibody [clone: 1/Stat1]
Supplier: Abcam
New Product
Certificates
About this item
Mouse monoclonal [1/Stat1] to STAT1.
Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: 1/Stat1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse
Frequently Bought Together