Order Entry
Italy
ContactUsLinkComponent
Anti-KAP1 (phospho S824) Rabbit Monoclonal Antibody [clone: BL-246-7B5]
Anti-KAP1 (phospho S824) Rabbit Monoclonal Antibody [clone: BL-246-7B5]
Catalog # ABCAAB243870-100
Supplier:  Abcam
undefined
Anti-KAP1 (phospho S824) Rabbit Monoclonal Antibody [clone: BL-246-7B5]
Catalog # ABCAAB243870-100
Supplier:  Abcam

Specifications

  • Size:
    100 µl
  • Pack type:
    Vial

Specifications

About this item

Rabbit monoclonal [BL-246-7B5] to KAP1 (phospho S824).

Type: Primary
Antigen: KAP1
Clonality: Monoclonal
Clone: BL-246-7B5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse