Anti-ENPP3/B10 Rabbit Monoclonal Antibody [clone: EPR27349-126]
: Abcam
New Product
Rabbit monoclonal [EPR27349-126] to ENPP3/B10.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ENPP3/B10
Clonality: Monoclonal
Clone: EPR27349-126
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
- Catalog No:
- ABCAAB313376-1ML
- ABCAAB313376-100UL
- ABCAAB313376-20UL
- Antigen symbol:
- ENPP3/B10
- ENPP3/B10
- ENPP3/B10
- Antigen name:
- CD203c
- CD203c
- CD203c
- Antigen synonyms:
- PDNP3|ENPP3|Ectonucleotide pyrophosphatase/phosphodiesterase family member 3|E-NPP 3|NPP3|Alkaline phosphodiesterase I|Dinucleoside polyphosphatase|Nucleotide diphosphatase|Nucleotide pyrophosphatase|Phosphodiesterase I beta|Phosphodiesterase I/nucleotide pyrophosphatase 3|NPPase|PD-Ibeta
- PDNP3|ENPP3|Ectonucleotide pyrophosphatase/phosphodiesterase family member 3|E-NPP 3|NPP3|Alkaline phosphodiesterase I|Dinucleoside polyphosphatase|Nucleotide diphosphatase|Nucleotide pyrophosphatase|Phosphodiesterase I beta|Phosphodiesterase I/nucleotide pyrophosphatase 3|NPPase|PD-Ibeta
- PDNP3|ENPP3|Ectonucleotide pyrophosphatase/phosphodiesterase family member 3|E-NPP 3|NPP3|Alkaline phosphodiesterase I|Dinucleoside polyphosphatase|Nucleotide diphosphatase|Nucleotide pyrophosphatase|Phosphodiesterase I beta|Phosphodiesterase I/nucleotide pyrophosphatase 3|NPPase|PD-Ibeta
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- EPR27349-126
- EPR27349-126
- EPR27349-126
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Liquid
- Molecular weight:
- 100124 Da
- 100124 Da
- 100124 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Protein A Affinity purification
- Storage temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Shipping temperature:
- 4 °C
- 4 °C
- 4 °C
- Tested applications:
- IHC-P
- IHC-P
- IHC-P
Frequently Bought Together