Anti-CD8 beta Rabbit Monoclonal Antibody [clone: EPR22331-54]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR22331-54] to CD8 beta.
Type: Primary
Antigen: CD8 beta
Clonality: Monoclonal
Clone: EPR22331-54
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Frequently Bought Together