Anti-Bag3 Rabbit Monoclonal Antibody [clone: EPR20207-58-4]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR20207-58-4] to Bag3 - BSA and Azide free.
Type: Primary
Antigen: Bag3
Clonality: Monoclonal
Clone: EPR20207-58-4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together