Order Entry
Italy
ContactUsLinkComponent
 
Anti-beta Actin Mouse Monoclonal Antibody [clone: AC-15] (HRP)
Catalog # ABCAAB49900
Supplier:  Abcam
undefined
Anti-beta Actin Mouse Monoclonal Antibody [clone: AC-15] (HRP)
Catalog # ABCAAB49900
Supplier:  Abcam

Specifications

  • Size:
    50 µl
  • Pack type:
    Vial

Specifications

About this item

HRP Mouse monoclonal [AC-15] to beta Actin.

Type: Primary
Antigen: beta Actin
Clonality: Monoclonal
Clone: AC-15
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse; Rat; Cow; Dog; Human; African Green Monkey; Hm