Anti-Histone H3 (methyl R63 + asymmetric di methyl R63 + symmetric di methyl R63) Rabbit Monoclonal Antibody [clone: EPR17722]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR17722] to Histone H3 (asymmetric di methyl R63, symmetric di methyl R63, methyl R63) - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR17722
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Frequently Bought Together