Order Entry
Italy
ContactUsLinkComponent
Anti-HLA Class I Mouse Monoclonal Antibody [clone: W6/32] (PE)
Anti-HLA Class I Mouse Monoclonal Antibody [clone: W6/32] (PE)
Catalog # ABCAAB33257-50
Supplier:  Abcam
undefined
Anti-HLA Class I Mouse Monoclonal Antibody [clone: W6/32] (PE)
Catalog # ABCAAB33257-50
Supplier:  Abcam

Specifications

  • Size:
    50 Tests
  • Pack type:
    Vial

Specifications

About this item

PE Mouse monoclonal [W6/32] to HLA Class I.

Type: Primary
Antigen: HLA Class I
Clonality: Monoclonal
Clone: W6/32
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human; Bb; Cat; Cow; Cynomolgus Monkey; Mcq; Monkey