Anti-Properdin/PFC Rabbit Monoclonal Antibody [clone: EPR25712-7]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR25712-7] to Properdin/PFC - BSA and Azide free (Capture).
Type: Primary
Antigen: Properdin/PFC
Clonality: Monoclonal
Clone: EPR25712-7
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together