Anti-Hsp70 Mouse Monoclonal Antibody [clone: 3A3]
Catalog # ABCAAB5439-1MG
Supplier: Abcam
New Product
Specifications
- Size:1 mg
- Pack type:Vial
Specifications
About this item
Mouse monoclonal [3A3] to Hsp70.
Type: Primary
Antigen: Hsp70
Clonality: Monoclonal
Clone: 3A3
Conjugation:
Epitope: Epitope mapping with a panel of Hsp 70 deletion mutants suggests that the epitope recognized is located between amino acids 504-617 of human Hsp 70, a region that has been shown to be involved in stress-induced nucleolar localization.
Host: Mouse
Isotype: IgG1
Reactivity: Mouse; Rat; Human