Order Entry
Italy
ContactUsLinkComponent
Anti-Leishmania A2 Mouse Monoclonal Antibody [clone: C9]
Anti-Leishmania A2 Mouse Monoclonal Antibody [clone: C9]
Catalog # ABCAAB150344-50
Supplier:  Abcam
undefined
Anti-Leishmania A2 Mouse Monoclonal Antibody [clone: C9]
Catalog # ABCAAB150344-50
Supplier:  Abcam

Specifications

  • Seq:
    1202
  • Size:
    50 µg
  • Pack type:
    Vial

Specifications

About this item

Mouse monoclonal [C9] to Leishmania A2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Leishmania A2
Clonality: Monoclonal
Clone: C9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Leish