Order Entry
Italy
ContactUsLinkComponent
Anti-beta Amyloid 1-40 Rabbit Monoclonal Antibody [clone: EPR23712-2]
Anti-beta Amyloid 1-40 Rabbit Monoclonal Antibody [clone: EPR23712-2]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-beta Amyloid 1-40 Rabbit Monoclonal Antibody [clone: EPR23712-2]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB254345-100
  • ABCAAB254345-1ML
  • ABCAAB254345-20UL
  • Antigen symbol:
  • beta Amyloid 1-40
  • beta Amyloid 1-40
  • beta Amyloid 1-40
  • Antigen name:
  • A4
  • A4
  • A4
  • Antigen synonyms:
  • AD1|APP|Amyloid-beta precursor protein|ABPP|APPI|Alzheimer disease amyloid A4 protein homolog|Alzheimer disease amyloid protein|Amyloid precursor protein|Amyloid-beta (A4) precursor protein|Amyloid-beta A4 protein|Cerebral vascular amyloid peptide|PreProtease nexin-II|CVAP|PN-II
  • AD1|APP|Amyloid-beta precursor protein|ABPP|APPI|Alzheimer disease amyloid A4 protein homolog|Alzheimer disease amyloid protein|Amyloid precursor protein|Amyloid-beta (A4) precursor protein|Amyloid-beta A4 protein|Cerebral vascular amyloid peptide|PreProtease nexin-II|CVAP|PN-II
  • AD1|APP|Amyloid-beta precursor protein|ABPP|APPI|Alzheimer disease amyloid A4 protein homolog|Alzheimer disease amyloid protein|Amyloid precursor protein|Amyloid-beta (A4) precursor protein|Amyloid-beta A4 protein|Cerebral vascular amyloid peptide|PreProtease nexin-II|CVAP|PN-II
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR23712-2
  • EPR23712-2
  • EPR23712-2
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Molecular weight:
  • 86943 Da
  • 86943 Da
  • 86943 Da
  • Purification:
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Storage temperature:
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
  • Shipping temperature:
  • 4 °C
  • 4 °C
  • 4 °C
  • Tested applications:
  • Dot, I-ELISA, IHC-P
  • Dot, I-ELISA, IHC-P
  • Dot, I-ELISA, IHC-P

Specifications

Frequently Bought Together