Order Entry
Italy
ContactUsLinkComponent
Anti-Complement C8a Rabbit Monoclonal Antibody [clone: EPR23646-142]
Anti-Complement C8a Rabbit Monoclonal Antibody [clone: EPR23646-142]
Supplier:  Abcam
Certificates
undefined
Anti-Complement C8a Rabbit Monoclonal Antibody [clone: EPR23646-142]
Supplier:  Abcam

Specifications

  • Catalog No:
  • ABCAAB285029-100
  • ABCAAB285029-1
  • Antigen symbol:
  • Complement C8a
  • Complement C8a
  • Antigen name:
  • Complement component C8 alpha chain
  • Complement component C8 alpha chain
  • Antigen synonyms:
  • Complement component 8 subunit alpha|C8A
  • Complement component 8 subunit alpha|C8A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR23646-142
  • EPR23646-142
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Molecular weight:
  • 65163 Da
  • 65163 Da
  • Purification:
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Storage temperature:
  • Store at 4 °C.
  • Store at 4 °C.
  • Shipping temperature:
  • 4 °C
  • 4 °C
  • Tested applications:
  • sELISA
  • sELISA

Specifications

Frequently Bought Together