Anti-Galectin 3 Rabbit Monoclonal Antibody [clone: EPR19244]
: Abcam
Rabbit monoclonal [EPR19244] to Galectin 3 - BSA and Azide free.
Type: Primary
Antigen: Galectin 3
Clonality: Monoclonal
Clone: EPR19244
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together