Anti-alpha smooth muscle Actin Rabbit Monoclonal Antibody [clone: EPR20062-77]
Nome del fornitore: Abcam
Certificati
Informazioni su questo articolo
Rabbit monoclonal [EPR20062-77] to alpha smooth muscle Actin - BSA and Azide free (Detector).
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: alpha smooth muscle Actin
Clonality: Monoclonal
Clone: EPR20062-77
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifiche
- Catalog No:
- ABCAAB245091-1
- ABCAAB245091-100
- Antigen symbol:
- alpha smooth muscle Actin
- alpha smooth muscle Actin
- Antigen name:
- ACTSA
- ACTSA
- Antigen synonyms:
- ACTVS|GIG46|ACTA2|Alpha-actin-2|Cell growth-inhibiting gene 46 protein
- ACTVS|GIG46|ACTA2|Alpha-actin-2|Cell growth-inhibiting gene 46 protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR20062-77
- EPR20062-77
- Form:
- Liquid
- Liquid
Specifiche
Frequently Bought Together