Anti-TRPV4 Rabbit Monoclonal Antibody [clone: EPR27068-39]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR27068-39] to TRPV4.
Type: Primary
Antigen: TRPV4
Clonality: Monoclonal
Clone: EPR27068-39
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Frequently Bought Together