Anti-TREM2 Rabbit Monoclonal Antibody [clone: EPR26210-1]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR26210-1] to TREM2.
Type: Primary
Antigen: TREM2
Clonality: Monoclonal
Clone: EPR26210-1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Frequently Bought Together