Order Entry
Italy
ContactUsLinkComponent
Anti-Engrailed and Invected Mouse Monoclonal Antibody [clone: 4D9]
Anti-Engrailed and Invected Mouse Monoclonal Antibody [clone: 4D9]
Catalog # ABCAAB12454-100
Supplier:  Abcam
undefined
Anti-Engrailed and Invected Mouse Monoclonal Antibody [clone: 4D9]
Catalog # ABCAAB12454-100
Supplier:  Abcam

Specifications

  • Seq:
    1286
  • Size:
    100 µg
  • Pack type:
    Vial

Specifications

About this item

Mouse monoclonal [4D9] to Engrailed and Invected.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Engrailed and Invected
Clonality: Monoclonal
Clone: 4D9
Conjugation:
Epitope: Epitope has been localized to residues 38-58 of the homeodomain.
Host: Mouse
Isotype: IgG1
Reactivity: Drosophila melanogaster