Order Entry
Italy
ContactUsLinkComponent
 
Anti-TCR V beta 3.1 Rabbit Polyclonal Antibody
  ABCAAB235044-100
 :  Abcam
undefined
Anti-TCR V beta 3.1 Rabbit Polyclonal Antibody
  ABCAAB235044-100
 :  Abcam

 

  • Size:
    100 µg
  • Pack type:
    Vial

 

 

Rabbit polyclonal to TCR V beta 3.1.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TCR V beta 3.1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human