Order Entry
Italy
ContactUsLinkComponent
Anti-CD10 Mouse Monoclonal Antibody [clone: MEM-78] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Anti-CD10 Mouse Monoclonal Antibody [clone: MEM-78] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Catalog # ABCAAB157307-50
Supplier:  Abcam
undefined
Anti-CD10 Mouse Monoclonal Antibody [clone: MEM-78] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Catalog # ABCAAB157307-50
Supplier:  Abcam

Specifications

  • Seq:
    0780
  • Size:
    50 Tests
  • Pack type:
    Vial

Specifications

About this item

PerCP/Cy5.5® Mouse monoclonal [MEM-78] to CD10, prediluted.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: MEM-78
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human