Order Entry
Italy
ContactUsLinkComponent
Anti-14-3-3 alpha + beta Rabbit Polyclonal Antibody
Anti-14-3-3 alpha + beta Rabbit Polyclonal Antibody
  ABCAAB237807-100
 :  Abcam
undefined
Anti-14-3-3 alpha + beta Rabbit Polyclonal Antibody
  ABCAAB237807-100
 :  Abcam

 

  • Seq:
    1286
  • Size:
    100 µg
  • Pack type:
    Vial

 

 

Rabbit polyclonal to 14-3-3 alpha + beta.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: 14-3-3 alpha + beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human, Zebrafish