Petri dishes, square
Supplier: Greiner Bio-One
Certificates
About this item
High grade PS. Square Petri dishes with exceptional optical clarity for microscopic analysis as well as heat resistance for use with hot agar.
Frequently Bought Together
High grade PS. Square Petri dishes with exceptional optical clarity for microscopic analysis as well as heat resistance for use with hot agar.