2,3-Dimethylbenzaldehyde ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5779-93-1
:
Formula:C₉H₁₀O
MDL Number:MFCD00798004
:134.18 g/mol
Boiling Pt:110°C
Flash Pt:102°C
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together