Order Entry
Italy
ContactUsLinkComponent
tert-Butyldimethylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Flammable
Corrosive
tert-Butyldimethylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
tert-Butyldimethylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together