

tert-Butyldimethylsilyl trifluoromethanesulphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69739-34-0
:
Formula:C₇H₁₅F₃O₃SSi
MDL Number:MFCD00000405
:264.34 g/mol
Boiling Pt:65…67 °C (12 mmHg)
Density:1.151
Flash Pt:36 °C (96 °F)
EINECS:274-102-0
UN:2920
ADR:8,II
:GHS05
:GHS02
Related Information
Frequently Bought Together