


tert-Butyldimethylsilyl trifluoromethanesulphonate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69739-34-0
:
Formula:C₇H₁₅F₃O₃SSi
MDL Number:MFCD00000405
:264.34 g/mol
Boiling Pt:65…67 °C (12 mmHg)
Flash Pt:36 °C
EINECS:274-102-0
UN:2920
ADR:8,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3850 to 1.3880 (20°C, 589 nm)
- Assay:≥97.5 %
Specification Test Results
Frequently Bought Together