Order Entry
Italy
ContactUsLinkComponent
Lithiumbis(trimethylsilyl)amide ∼26% ∼1.3 mol/L in THF
Flammable
Corrosive
Irritant
Health
Lithiumbis(trimethylsilyl)amide ∼26% ∼1.3 mol/L in THF
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Lithiumbis(trimethylsilyl)amide ∼26% ∼1.3 mol/L in THF
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green clear liquid to cloudy liquid
  • Concentration (Neutralization titration):
    23.0 to 29.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together