
4-(Trifluoromethylthio)phenol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:461-84-7
:
Formula:C₇H₅F₃OS
MDL Number:MFCD00036035
:194.18 g/mol
Boiling Pt:78°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
Specification Test Results
Frequently Bought Together