
Bis(tri-tert-butylphosphine)palladium(0) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:53199-31-8
:
Formula:C₂₄H₅₄P₂Pd
MDL Number:MFCD03094580
:511.05 g/mol
Melting Pt:258…272 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Off-white to orange-brown Powder or crystals
- Infrared spectrum:Conforms
- Assay:≥97.5 % (ex Pd)
Specification Test Results
Frequently Bought Together