
Nicotinoyl chloride hydrochloride ≥95.0% (by titrimetric analysis)
: TCI
:20260-53-1
:
:C₆H₅Cl₂NO
:MFCD00012820
:178.02 g/mol
:243-648-1
:3261
:8,III
:GHS05
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 95.0 %
- Melting point:152.0 to 157.0 deg-C
Frequently Bought Together